Learn More
Biomatik Corporation Rat Alkaline Sphingomyelinase (Alk-Smase) ELISA Kit
Supplier:  Biomatik Corporation EKU02253
Encompass_Preferred
Rat Alkaline Sphingomyelinase (Alk-Smase) ELISA Kit for detecting Alk-Smase in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.047ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5EZ72. Gene ID: 303729. Target Synonyms: ENPP7; E-NPP 7; NPP-7; Ectonucleotide Pyrophosphatase/Phosphodiesterase 7; Sphingomyelin Phosphodiesterase; Intestinal alkaline sphingomyelinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9144