Learn More
American Research Products Inc Rat alpha 1 microglobulin biku
Catalog No. 5000328457
Encompass
Rat alpha-1-microglobulin/bikunin precursor (AMBP) ELISA kit from CUSABIO Detection Range 1 563 ng/mL-100 ng/mL Sensitivity 0 649 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28457 Supplier American Research Products Inc Supplier No. CSBEL001654RA
Rat alpha-1-microglobulin/bikunin precursor (AMBP) ELISA kit from CUSABIO Detection Range 1 563 ng/mL-100 ng/mL Sensitivity 0 649 ng/mL Sample serum plasma tissue homogenates Method Competitive