Learn More
Abcam Rat alpha 2 Macroglobulin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1577301X96TESTS
Encompass_Preferred
Rat alpha 2 Macroglobulin ELISA Kit ab157730 is used to quantify Rat alpha 2 Macroglobulin with a sensitivity of 1.795 ng/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1952812