Learn More
American Research Products Inc Rat Alpha fetoprotein AFP ELI
Catalog No. 5000329690
Encompass
Rat Alpha-fetoprotein AFP ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29690 Supplier American Research Products Inc Supplier No. CSBE08281R
Rat Alpha-fetoprotein AFP ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich