Learn More
Biomatik Corporation Rat Alpha-2-Glycoprotein 1, Zinc Binding (aZGP1) ELISA Kit
Supplier:  Biomatik Corporation EKU09412
Encompass_Preferred
Rat Alpha-2-Glycoprotein 1, Zinc Binding (aZGP1) ELISA Kit for detecting aZGP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000ng/ml. Sensitivity: 7.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63678. Target Synonyms: ZA2G; ZAG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4425