Learn More
Biomatik Corporation Rat Alpha-2-Macroglobulin (a2M) ELISA Kit
Supplier:  Biomatik Corporation EKU10625
Encompass_Preferred
Rat Alpha-2-Macroglobulin (a2M) ELISA Kit for detecting a2M in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000ng/ml. Sensitivity: 58.4ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P06238. Gene ID: 24153. Target Synonyms: A2MG; A2mg; CPAMD5; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7346