Learn More
Biomatik Corporation Rat Alpha-Melanocyte Stimulating Hormone (aMSH) ELISA Kit
Supplier:  Biomatik Corporation EKU08457
Encompass_Preferred
Rat Alpha-Melanocyte Stimulating Hormone (aMSH) ELISA Kit for detecting aMSH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 51.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01194. Target Synonyms: α-MSH; Intermedins; Alpha-Melanotropin, Alpha-Melanocortin; Alpha-Intermedin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6270