Learn More
Biomatik Corporation Rat Aminolevulinate Delta Dehydratase (ALAD) ELISA Kit
Supplier:  Biomatik Corporation EKU02315
Encompass_Preferred
Rat Aminolevulinate Delta Dehydratase (ALAD) ELISA Kit for detecting ALAD in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06214. Gene ID: 25374. Target Synonyms: ALADH; PBGS; Delta-Aminolevulinic Acid Dehydratase; Porphobilinogen Synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9141