Learn More
BIOHIPPO Rat AMN Microsample
SDP
Catalog No. 5000314865
Encompass
Rat AMN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of AMN in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 21 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat AMN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat AMN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat AMN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14865 Supplier BIOHIPPO Supplier No. BHE15210830
Rat AMN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of AMN in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 21 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat AMN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat AMN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat AMN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color