Learn More
Biomatik Corporation Rat Amylase, Alpha 2A (AMY2A) ELISA Kit
Supplier:  Biomatik Corporation EKU11696
Encompass_Preferred
Rat Amylase, Alpha 2A (AMY2A) ELISA Kit for detecting AMY2A in serum, plasma, saliva, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G3V844. Target Synonyms: AMY2; AMY2B; PA; 1,4-alpha-D-glucan glucanohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7653