Learn More
Biomatik Corporation Rat Amyloid Precursor Protein (APP) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10517
Encompass_Preferred
Rat Amyloid Precursor Protein (APP) Wide-range ELISA Kit for detecting APP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.48ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08592. Gene ID: 54226. Target Synonyms: AAA; ABeta; ABPP; AD1; APPI; CVAP; PN2; CTFgamma; Peptidase Nexin-II Alzheimer Disease; Peptidase Nexin-II; Amyloid Beta (A4) Precursor Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8331