Learn More
American Research Products Inc Rat Androgen receptor AR ELI
Catalog No. 5000327763
Encompass
Rat Androgen receptor (AR) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27763 Supplier American Research Products Inc Supplier No. CSBEL001975RA
Rat Androgen receptor (AR) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich