Learn More
Raybiotech Inc Rat Angiopoietin-1 ELISA Kit
Supplier:  Raybiotech Inc ELRANGIOPOIETIN11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.4 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects rat angiopoietin-1 and human angiopoietin-1. Other species not determined.
Catalog No. 50-195-0215