Learn More
Biomatik Corporation Rat Angiopoietin Like Protein 6 (ANGPTL6) ELISA Kit
Supplier:  Biomatik Corporation EKU02378
Encompass_Preferred
Rat Angiopoietin Like Protein 6 (ANGPTL6) ELISA Kit for detecting ANGPTL6 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.262ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B2RYM1. Target Synonyms: ARP5; AGF; Angiopoietin-Related Protein 5; Angiopoietin-related growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5254