Learn More
American Research Products Inc Rat Angiopoietin related prote
Catalog No. 5000327117
Encompass
Rat Angiopoietin-related protein 4 (ANGPTL4) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates bronchoalveolar lavage fluid urine Method Sandwich
Catalog No. 50-003-27117 Supplier American Research Products Inc Supplier No. CSBEL001712RA
Rat Angiopoietin-related protein 4 (ANGPTL4) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates bronchoalveolar lavage fluid urine Method Sandwich