Learn More
American Research Products Inc Rat angiotensin 1 7 Ang1 7 E
Catalog No. 5000330254
Encompass
Rat angiotensin 1-7 (Ang1-7) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30254 Supplier American Research Products Inc Supplier No. CSBE14241R
Rat angiotensin 1-7 (Ang1-7) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich