Learn More
Medchemexpress LLC Rat Anti-Mouse CD86 Antibody (FITC)(YA3447) | 50 TESTS
SDP
Catalog No. 5000444815
Encompass_Preferred
This antibody is derived from Rat and reacts with Mouse CD86. It is suitable for Flow Cytometry (FC) applications. The product is supplied in PBS with 0.05% Proclin300 and 1% BSA.
  • Derived from Rat
  • Reacts with Mouse CD86
  • Suitable for Flow Cytometry (FC)
  • Stored at 2-8°C for 1 year
  • Protect from light, do not freeze
  • Supplied in PBS with 0.05% Proclin300, 1% BSA
  • Concentration: 5 μL per test
Catalog No. 50-004-44815 Supplier Medchemexpress LLC Supplier No. HYP83710M50T
This antibody is derived from Rat and reacts with Mouse CD86. It is suitable for Flow Cytometry (FC) applications. The product is supplied in PBS with 0.05% Proclin300 and 1% BSA.
  • Derived from Rat
  • Reacts with Mouse CD86
  • Suitable for Flow Cytometry (FC)
  • Stored at 2-8°C for 1 year
  • Protect from light, do not freeze
  • Supplied in PBS with 0.05% Proclin300, 1% BSA
  • Concentration: 5 μL per test