Learn More
AbD Serotec RAT ANTI-MOUSE F4/80 ANTIGEN
Catalog No. NC9998488
Encompass
Rat Anti Mouse, Specificity: F4/80 Antigen, Product Type: Monoclonal Antibody, Clone: CI:A3-1, Isotype: IgG2b, Quantity/Volume: 0.1mg, Phosphate buffered saline pH 7.4, Preservatives Stabilisers: 0.09% Sodium Azide
Catalog No. NC9998488 Supplier AbD Serotec Supplier No. MCA497GA
Rat Anti Mouse, Specificity: F4/80 Antigen, Product Type: Monoclonal Antibody, Clone: CI:A3-1, Isotype: IgG2b, Quantity/Volume: 0.1mg, Phosphate buffered saline pH 7.4, Preservatives Stabilisers: 0.09% Sodium Azide