Learn More
AbD Serotec RAT ANTI MOUSE F480 ANTIGEN
Catalog No. NC9256507
Encompass
Antibody; Monoclonal; Rat Anti Mouse F4/80 Antigen, Clone: CI: A3-1 (F4/80); ABD Seritec; IgG Conc.: 1mg/mL; Isotype: IgG2b (Rat); Applications: C, EM, F, IF, IP, P *, R, RE, WB
Catalog No. NC9256507 Supplier AbD Serotec Supplier No. MCA497R
Antibody; Monoclonal; Rat Anti Mouse F4/80 Antigen, Clone: CI: A3-1 (F4/80); ABD Seritec; IgG Conc.: 1mg/mL; Isotype: IgG2b (Rat); Applications: C, EM, F, IF, IP, P *, R, RE, WB