Learn More
Rat anti-Mouse IgD, Clone: 11-26c.2a, FITC, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The 11-26c.2a antibody reacts with mouse immunoglobulin D of all Igh-C haplotypes (e.g., IgDa, IgDb, IgDe), and it does not react with other immunoglobulin isotypes. Although 11-26c.2a mAb binds membrane IgD expressed on the splenic B-cell surface with hi
Host Species: Rat
Clone: 11-26c.2a
Isotype: IgG2a, κ
Species Reactivity: Mouse
Flow Cytometry
Specifications
Specifications
| Antigen | Rat anti-Mouse IgD |
| Classification | Monoclonal |
| Clone | 11-26C.2A |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Rat |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.