Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgE, Clone: R35-118, Secondary Antibody, BD

Catalog No. BDB553419 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553419 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553419 Supplier BD Biosciences Supplier No. 553419
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The R35-118 clone has been reported to react specifically with mouse IgE of Igh-C[a] and Igh-C[b] haplotypes. It has been reported not to react with other Ig isotypes. This antibody is routinely tested by ELISA analysis.

Host Species: Rat
Clone: R35-118
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled mouse IgE

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse IgE
Classification Monoclonal
Clone R35-118
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Rat
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.