Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgE, Clone: R35-72, biotin, Secondary Antibody, BD

Catalog No. BDB553414 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553414 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553414 Supplier BD Biosciences Supplier No. 553414
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The rat anti-mouse IgE antibody (clone R35-72) reacts specifically with mouse IgE of the Igh-C [a] and Igh-C [b] haplotypes. It has been reported not to react with other Ig isotypes. Detection with the rat anti-mouse IgE antibody (clone R35-72) of surface

Host Species: Rat
Clone: R35-72
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Mouse IgE (pooled)

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse IgE
Classification Monoclonal
Clone R35-72
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Rat
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.