Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgE Heavy Chain Secondary Antibody (LO-ME-3), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB554847
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554847 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554847 Supplier R&D Systems Supplier No. NBP268431G

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

IgE Heavy Chain Monoclonal specifically detects IgE Heavy Chain in Mouse samples. It is validated for ELISA, Western Blot (Negative).

Specifications

Antigen IgE Heavy Chain
Applications ELISA, Western Blot (Negative)
Classification Monoclonal
Clone LO-ME-3
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon
Host Species Rat
Immunogen Purified IgE from BALB/c mice
Purification Method Affinity purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.