Learn More
Rat anti-Mouse IgE Heavy Chain Secondary Antibody (LO-ME-3), PerCP, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgE Heavy Chain |
| Applications | ELISA, Western Blot (Negative) |
| Classification | Monoclonal |
| Clone | LO-ME-3 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon |
| Host Species | Rat |
| Immunogen | Purified IgE from BALB/c mice |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.