Learn More
Rat anti-Mouse IgG Heavy Chain Secondary Antibody (LO-MG-7), Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG Heavy Chain |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | LO-MG-7 |
| Concentration | 1.0mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Frozen |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Host Species | Rat |
| Immunogen | Synthetic peptide encompassing the unique 20 N-terminal amino acid sequence of RTN1-C |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.