Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Anti-Mouse IgG1, AKP, Clone: X56, BD

Catalog No. BDB557272 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557272 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557272 Supplier BD Biosciences Supplier No. 557272
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The X56 antibody reacts specifically with mouse IgG1 of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: X56
Isotype: IgG1 κ
Species Reactivity: Mouse

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1
Classification Monoclonal
Clone X56
Conjugate Alkaline Phosphatase (AP)
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.