Learn More
Rat anti-Mouse IgG1, Clone: A85-1, biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The A85-1 antibody reacts specifically with mouse IgG1 of Igh-Ca and Igh-Cb haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with A85-1 mAb. A suspension of pooled mouse Ig
Host Species: Rat
Clone: A85-1
Isotype: IgG1, κ
Species Reactivity: Mouse
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Rat anti-Mouse IgG1 |
| Classification | Monoclonal |
| Clone | A85-1 |
| Conjugate | Biotin |
| Host Species | Rat |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Target Species | Mouse |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.