Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgG1, Secondary Antibody, BD

Catalog No. BDB562580 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562580 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562580 Supplier BD Biosciences Supplier No. 562580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The A85-1 antibody reacts specifically with mouse IgG1 of Igh-Ca and Igh-Cb haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with the A85-1 monoclonal antibody. A suspension of pooled mouse IgG1 was used as the source of immunogen.

Host Species: Rat
Clone: A85-1
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse IgG1

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG1
Classification Monoclonal
Clone A85-1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Host Species Rat
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.