Learn More
Rat anti-Mouse IgG2a/2b, Clone: R2-40, FITC, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The R2-40 antibody recognizes a common epitope, probably located in the CH1 domain, shared by mouse IgG2a, and IgG2b, of Igh-Ca and Igh-Cb haplotypes. It does not react with other Ig isotypes. This antibody is routinely tested by ELISA analysis.
Host Species: Rat
Clone: R2-40
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Rat anti-Mouse IgG2a/2b |
| Classification | Monoclonal |
| Clone | R2-40 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Rat |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.