Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgG2a/2b, Clone: R2-40, Secondary Antibody, BD

Catalog No. BDB553397 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553397 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553397 Supplier BD Biosciences Supplier No. 553397
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The R2-40 antibody recognizes a common epitope, probably located in the CH1 domain, shared by mouse IgG2a, and IgG2b, of Igh-Ca and Igh-Cb haplotypes. It does not react with other Ig isotypes. This antibody is routinely tested by ELISA analysis.

Host Species: Rat
Clone: R2-40
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse IgG2a/2b
Classification Monoclonal
Clone R2-40
Concentration 0.5mg/mL
Host Species Rat
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.