Learn More
Rat anti-Mouse IgG2a, Clone: R19-15, biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The R19-15 antibody recognizes an epitope in the CH3 domain of mouse IgG2a, with strong reactivity to the Igh-I[a] allotype and weaker reactivity to Igh-I[b]. It does not react with other Ig isotypes. Molecular genetic analyses suggest that the Igh-I[b] a
Host Species: Rat
Clone: R19-15
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled BALB/c and C57BL/6 mouse Ig
ELISA
Specifications
Specifications
| Antigen | Rat anti-Mouse IgG2a |
| Classification | Monoclonal |
| Clone | R19-15 |
| Conjugate | Biotin |
| Host Species | Rat |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Target Species | Mouse |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.