Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat anti-Mouse IgG2a Secondary Antibody, Biotin

Catalog No. 046240
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-624-0 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-624-0 Supplier Invitrogen™ Supplier No. 046240
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Secondary Antibody

This monoclonal Rat anti-Mouse IgG2a reacts with mouse IgG2a and exhibits no reactivity to the other mouse IgG subclasses, IgA, IgM or light chains.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG2a
Applications ELISA
Classification Monoclonal
Clone LO-MG2a-3
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 1% BSA, 40% glycerol and 0.1% sodium azide; pH 7.4
Host Species Rat
Immunogen Mouse IgG2a.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.