Learn More
Rat anti-Mouse IgG2b, Clone: R12-3, FITC, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The R12-3 antibody recognizes an epitope in the CH3 domain of mouse IgG2b of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with R12-3 mAb. This ant
Host Species: Rat
Clone: R12-3
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Rat anti-Mouse IgG2b |
| Classification | Monoclonal |
| Clone | R12-3 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Rat |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.