Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgG2b, Clone: R9-91, Secondary Antibody, BD

Catalog No. BDB553396 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553396 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553396 Supplier BD Biosciences Supplier No. 553396

missing translation for 'validMainframeMsg'

The R9-91 antibody recognizes an epitope in the CH3 domain of mouse IgG2b of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with R9-91 mAb

Host Species: Rat
Clone: R9-91
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse IgG2b
Classification Monoclonal
Clone R9-91
Concentration 0.5mg/mL
Host Species Rat
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.