Learn More
Rat anti-Mouse IgG2b Heavy Chain Secondary Antibody (LO-MG2b-2), Allophycocyanin, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG2b Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LO-MG2b-2 |
| Conjugate | APC |
| Formulation | PBS |
| Host Species | Rat |
| Immunogen | Purified mouse IgG2b |
| Purification Method | Protein A purified |
| Quantity | 0.25 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.