Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat anti-Mouse IgG2b Secondary Antibody, FITC

Catalog No. 01672172 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-172 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-172 Supplier Invitrogen™ Supplier No. SA135649
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Secondary Antibody

Mouse Gamma 2b Heavy Chain of Immunoglobulin (determined by immunodot). positively tested on BALB/c and C57BL/6 mouse sera. The specificity of every rat monoclonal antibody anti-mouse IgG subclasses is determined in a range of optimal concentrations. Increasing the first or the second antibody at an unnecessary too high concentration can induce possible cross-reactions with other mouse IgG subclasses. Avidity on IgG2b: 1 * 1010 M-1. No cross-reactivity with chicken, rabbit, goat, sheep, bovine, horse, dog, swine, baboon IgG and human IgG or IgM (ELISA test). Available unconjugated MON5055, FITC MON5055F, Biotin MON5055B, PE MON5055P.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG2b
Applications ELISA, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone LO-MG2b-2
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.1% sodium azide
Host Species Rat
Immunogen Immunoglobulins.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C. Store in the dark.
Product Type Secondary Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.