Learn More
Description
The R2-38 antibody reacts specifically with mouse IgG3 of all strains. It does not react with other Ig isotypes nor does it react with IgG3 of M. spretus. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with R2-38 mAb.
Host Species: Rat
Clone: R2-38
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Ig from BALB/c Mouse Plasmacytomas FLOPC 21 (IgG3, κ) and Y 5606 (IgG3, λ)
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Rat anti-Mouse IgG3 |
| Classification | Monoclonal |
| Clone | R2-38 |
| Concentration | 0.5mg/mL |
| Host Species | Rat |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.