Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgM, Clone: II/41, Secondary Antibody, BD

Catalog No. BDB553435 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553435 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553435 Supplier BD Biosciences Supplier No. 553435
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The II/41 clone has been reported to react specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It has been reported not to react with other Ig isotypes. In addition, the II/41 clone has been reported not to stimulate B-cell proliferation. Thi

Host Species: Rat
Clone: II/41
Isotype: IgG2a, κ
Species Reactivity: Mouse

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse IgM
Classification Monoclonal
Clone II/41
Concentration 0.5mg/mL
Host Species Rat
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.