Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Anti-Mouse IgM, Clone: R6-60.2, PE, Secondary Antibody, BD

Catalog No. BDB552867 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552867 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552867 Supplier BD Biosciences Supplier No. 552867
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The R6-60.2 antibody reacts specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. R6-60.2 antibody has not been shown to stimulate B-cell proliferation. The antibody is conjugated to BD Horizon™

Host Species: Rat
Clone: R6-60.2
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat Anti-Mouse IgM
Classification Monoclonal
Clone R6-60.2
Concentration 0.2mg/mL
Conjugate PE
Host Species Rat
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.