Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Immunoglobulin kappa light chainRat anti-Mouse, Clone: 187.1, Secondary Antibody, MilliporeSigma™

Catalog No. MAB10758MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10758MI 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10758MI Supplier MilliporeSigma Supplier No. MAB10758

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Anti-mouse Immunoglobulin kappa light chain Antibody, clone 187.1, Rat is an antibody against mouse Immunoglobulin kappa light chain for use in ELISA, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Immunoglobulin kappa light chain
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 187.1
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Rat
Immunogen Mouse IgG2b, κ secreted by MPC-11 plasmacytoma
Purification Method Protein G
Quantity 100 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.