Learn More
Biomatik Corporation Rat Actin Alpha 1, Cardiac Muscle (Anti-ACTC1) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11957
Encompass_Preferred
Rat Actin Alpha 1, Cardiac Muscle (Anti-ACTC1) Anti-antigen ELISA Kit for detecting Anti-ACTC1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Target Synonyms: ACTC; CMD1R; CMD1-R; ACT-a1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8060