Learn More
Biomatik Corporation Rat Cardiac Troponin I (cTnI) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11894
Encompass_Preferred
Rat Cardiac Troponin I (cTnI) Anti-antigen ELISA Kit for detecting cTnI in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.14ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P23693. Gene ID: 29248. Target Synonyms: TNNI3; TNNC1; CMH7; C-TnI; Troponin I Type 3; Troponin I Type 3, Cardiac. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5470