Learn More
Biomatik Corporation Rat Anti-Mullerian Hormone (AMH) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10518
Encompass_Preferred
Rat Anti-Mullerian Hormone (AMH) Wide-range ELISA Kit for detecting AMH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 50.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49000. Gene ID: 25378. Target Synonyms: MIF; MIH; MIS; Müllerian Inhibiting Factor; Müllerian Inhibiting Hormone; Müllerian Inhibiting Substance. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5273