Learn More
Biomatik Corporation Rat Myosin Heavy Chain 7B, Cardiac Muscle Beta (MYH7B) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12098
Encompass_Preferred
Rat Myosin Heavy Chain 7B, Cardiac Muscle Beta (MYH7B) Anti-antigen ELISA Kit for detecting MYH7B in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.27ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Target Synonyms: MYH14; MHC14; Antigen MLAA-21; Myosin cardiac muscle beta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8030