Learn More
Biomatik Corporation Rat Thyroid Peroxidase (TPO) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11905
Encompass_Preferred
Rat Thyroid Peroxidase (TPO) Anti-antigen ELISA Kit for detecting TPO in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.12ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P14650. Target Synonyms: TPX; thyroperoxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6089