Learn More
Lonza Walkersville Rat Aortic Smooth Muscle Cells
Catalog No. 5000496433
Encompass
Cryopreserved Rat AoSMCs are isolated from adult Sprague Dawley rats These cells are characterized and test 95% positive for smooth muscle actin Use these cells for your angiogenesis artherosclerosis and cardiovascular research Cells can also be utilized to cross-compare your studies with Clonetics Primary Human Aortic Smooth Muscle Cells
Catalog No. 50-004-96433 Supplier Lonza Walkersville Supplier No. RASM580
Cryopreserved Rat AoSMCs are isolated from adult Sprague Dawley rats These cells are characterized and test 95% positive for smooth muscle actin Use these cells for your angiogenesis artherosclerosis and cardiovascular research Cells can also be utilized to cross-compare your studies with Clonetics Primary Human Aortic Smooth Muscle Cells