Learn More
Biomatik Corporation Rat Apelin (APLN) ELISA Kit
Supplier:  Biomatik Corporation EKU10651
Encompass_Preferred
Rat Apelin (APLN) ELISA Kit for detecting APLN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8,000pg/ml. Sensitivity: 51pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9R0R3. Gene ID: 58812. Target Synonyms: XNPEP2; APEL; APJ endogenous ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8318