Learn More
Sino Biological Rat APOH Protein 50ug (Sino Biological)
Supplier:  Sino Biological 201991T36100
Encompass_Preferred
A DNA sequence encoding the rat Apoh (NP_001009626.1) (Met1-Cys345) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-4052