Learn More
Biomatik Corporation Rat Apolipoprotein A5 (APOA5) ELISA Kit
Supplier:  Biomatik Corporation EKU02497
Encompass_Preferred
Rat Apolipoprotein A5 (APOA5) ELISA Kit for detecting APOA5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QUH3. Gene ID: 140638. Target Synonyms: Apo-A5; APOA-V; APOAV; RAP3; Regeneration-associated protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9126