Learn More
Biomatik Corporation Rat Apolipoprotein C1 (APOC1) ELISA Kit
Supplier:  Biomatik Corporation EKU08992
Encompass_Preferred
Rat Apolipoprotein C1 (APOC1) ELISA Kit for detecting APOC1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19939. Gene ID: 25292. Target Synonyms: Apo-C1; Truncated apolipoprotein C-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5895